Anti-TBC1D3 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB126411-100EA 0 SEK
BIRBORB126411-100 BIRBORB126411-200
Anti-TBC1D3 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TBC1D3 (TBC1 domain family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR