Anti-TBL2 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126412-100EA 0 SEK
BIRBORB126412-100 BIRBORB126412-200
Anti-TBL2 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TBL2 (transducin (beta)-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human