Anti-TBX1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126414-100EA 0 SEK
BIRBORB126414-100 BIRBORB126414-200
Anti-TBX1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TBX1 (T-box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human