Anti-TBX10 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126415-200EA 0 SEK
BIRBORB126415-200 BIRBORB126415-100
Anti-TBX10 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TBX10 (T-box 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human