Anti-TCEAL3/5/6 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126417-200EA 0 SEK
BIRBORB126417-200 BIRBORB126417-100
Anti-TCEAL3/5/6 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TCEAL3/5/6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human