Anti-TEP1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126421-200EA 0 SEK
BIRBORB126421-200 BIRBORB126421-100
Anti-TEP1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TEP1 (telomerase-associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human