Anti-THBD Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126425-200EA 0 SEK
BIRBORB126425-200 BIRBORB126425-100
Anti-THBD Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: THBD (thrombomodulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human