Anti-TLE2 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126433-100EA 0 SEK
BIRBORB126433-100 BIRBORB126433-200
Anti-TLE2 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TLE2 (transducin-like enhancer of split 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human