Anti-TNFSF10 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126440-200EA 0 SEK
BIRBORB126440-200 BIRBORB126440-100
Anti-TNFSF10 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TNFSF10 (tumor necrosis factor (ligand) superfamily, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human