Anti-TRIM25 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126458-100EA 0 SEK
BIRBORB126458-100 BIRBORB126458-200
Anti-TRIM25 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TRIM25 (tripartite motif containing 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human