Anti-TSEN54 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126459-100EA 0 SEK
BIRBORB126459-100 BIRBORB126459-200
Anti-TSEN54 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TSEN54 (TSEN54 tRNA splicing endonuclease subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human