Anti-TUBGCP6 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126462-100EA 0 SEK
BIRBORB126462-100 BIRBORB126462-200
Anti-TUBGCP6 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: TUBGCP6 (tubulin, gamma complex associated protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human