Anti-UBA7 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126469-200EA 0 SEK
BIRBORB126469-200 BIRBORB126469-100
Anti-UBA7 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: UBA7 (ubiquitin-like modifier activating enzyme 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human