Anti-UBAC1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126470-200EA 0 SEK
BIRBORB126470-200 BIRBORB126470-100
Anti-UBAC1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: UBAC1 (UBA domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human