Anti-UBXN11 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126473-100EA 0 SEK
BIRBORB126473-100 BIRBORB126473-200
Anti-UBXN11 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: UBXN11 (UBX domain protein 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human