Anti-URB1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126475-100EA 0 SEK
BIRBORB126475-100 BIRBORB126475-200
Anti-URB1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: URB1 (URB1 ribosome biogenesis 1 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human