Anti-TP53 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB137499-200EA 0 SEK
BIRBORB137499-200
Anti-TP53 Rabbit Polyclonal Antibody
Antikroppar
Immunofluorescence: 1:10-1:50; Immunochemistry: 10 ?g/ml; Western Blot: 1:1000

Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR