Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-ADAMTS19 Rabbit polyclonal antibody unconjugated

Leverantör: Biorbyt
Immunochemistry: 10 μg/ml; Western Blot: 1:50-1:100

Type: Primary
Antigen: ADAMTS19 (ADAM metallopeptidase with thrombospondin type 1 motif, 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:



 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
200 µl 200 µl BIRBORB137517-200
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol ADAMTS19
Antigen name ADAM metallopeptidase with thrombospondin type 1 motif, 19
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen ADAMTS19 antibody was raised against kLH conjugated synthetic peptide selected from the center region of human ADAMTS19.
Rening purified by immunoaffinity chromatography
Storage buffer PBS with 0,09% (w/v) sodium azide
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal