Anti-IGFBP3 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB137534-200EA 0 SEK
BIRBORB137534-200
Anti-IGFBP3 Rabbit Polyclonal Antibody
Antikroppar
Immunofluorescence: 1:10-1:50; Immunochemistry: 5 ?g/ml; Western Blot: 1:100-1:500

Type: Primary
Antigen: IGFBP3 (insulin-like growth factor binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR