Anti-IL1RL2 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB137540-200EA 0 SEK
BIRBORB137540-200
Anti-IL1RL2 Rabbit Polyclonal Antibody
Antikroppar
Immunochemistry: 5 ?g/ml; Western Blot: 1:100-1:500

Type: Primary
Antigen: IL1RL2 (interleukin 1 receptor-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR