Anti-INS Mouse Monoclonal Antibody

Leverantör: Biorbyt
BIRBORB137542-50EA 0 SEK
BIRBORB137542-50
Anti-INS Mouse Monoclonal Antibody
Antikroppar
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR