Anti-CDK2AP1 Rabbit polyclonal antibody

Leverantör: Biorbyt
BIRBORB137575-50EA 0 SEK
BIRBORB137575-50
Anti-CDK2AP1 Rabbit polyclonal antibody
Antikroppar
ELISA: 1:1000; Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CDK2AP1 (cyclin-dependent kinase 2 associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR