Anti-HARS Rabbit polyclonal antibody

Leverantör: Biorbyt
BIRBORB137580-50EA 0 SEK
BIRBORB137580-50
Anti-HARS Rabbit polyclonal antibody
Antikroppar
ELISA: 1:20000; Immunofluorescence: 1:100-1:500; Immunochemistry: 20 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: HARS (histidyl-tRNA synthetase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR