Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-MYF5 Rabbit polyclonal antibody unconjugated

Leverantör: Biorbyt
ELISA: 1:40000; Immunochemistry: 30 μg/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: MYF5 (myogenic factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:



 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG BIRBORB137581-50
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol MYF5
Antigen name Myogenic factor 5
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen MYF5/MYF 5 antibody was raised against synthetic peptide from human MYF5.
Rening purified by immunoaffinity chromatography
Storage buffer PBS (without Mg2+, Ca2+), pH 7,4, 150 mM sodium chloride, 0,02% sodium azide, 50% glycerol
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal