Anti-GTF3C3 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB137596-50EA 0 SEK
BIRBORB137596-50
Anti-GTF3C3 Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:1000; Immunochemistry: 20 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: GTF3C3 (general transcription factor IIIC, polypeptide 3, 102kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR