Anti-HSPH1 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96486-50EA 0 SEK
BIRBORB96486-50
Anti-HSPH1 Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:10000; Immunofluorescence: 1:100-1:500; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: HSPH1 (heat shock 105kDa/110kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR