Anti-Cystatin A Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96491-50EA 0 SEK
BIRBORB96491-50
Anti-Cystatin A Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:5000; Immunofluorescence: 1:100-1:500; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: Cystatin A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR