Anti-MAPK3 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96509-50EA 0 SEK
BIRBORB96509-50
Anti-MAPK3 Rabbit Polyclonal Antibody
Antikroppar
Immunochemistry: 1:100; Western Blot: 1:5000

Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR