Anti-BRI3BP Rabbit polyclonal antibody unconjugated

Leverantör: Biorbyt
BIRBORB96524-50EA 0 SEK
BIRBORB96524-50
Anti-BRI3BP Rabbit polyclonal antibody unconjugated
Antikroppar
ELISA: 1:20000; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: BRI3BP (BRI3 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR