Anti-HDAC1 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96531-50EA 0 SEK
BIRBORB96531-50
Anti-HDAC1 Rabbit Polyclonal Antibody
Antikroppar
Immunochemistry: 5-7.5 ?g/ml; Immunoprecipitation: 10-20 ?g/ml; Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR