Anti-FGFR4 Mouse Monoclonal Antibody

Leverantör: Biorbyt
BIRBORB96534-50EA 0 SEK
BIRBORB96534-50
Anti-FGFR4 Mouse Monoclonal Antibody
Antikroppar
  • ELISA: 1:1000; Immunochemistry: 1:50; Western Blot: 1:2000

Type: Primary
Antigen: FGFR4 (fibroblast growth factor receptor 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR