Anti-H3F3B Mouse Monoclonal Antibody

Leverantör: Biorbyt
BIRBORB96535-50EA 6950 SEK
BIRBORB96535-50
Anti-H3F3B Mouse Monoclonal Antibody
Antikroppar
  • Immunofluorescence: 10 μg/ml; Immunochemistry: 15 μg/ml

Type: Primary
Antigen: H3F3B (H3 histone, family 3B (H3.3B))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR