Anti-PSAT1 Mouse Monoclonal Antibody

Leverantör: Biorbyt
BIRBORB96546-50EA 0 SEK
BIRBORB96546-50
Anti-PSAT1 Mouse Monoclonal Antibody
Antikroppar
Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR