Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-PSAT1 Mouse monoclonal antibody unconjugated

Leverantör: Biorbyt
Immunochemistry: 10 μg/ml

Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:



 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG BIRBORB96548-50
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol PSAT1
Antigen name Phosphoserine Aminotransferase 1
Conjugation Unconjugated
Clonality Monoclonal
Antikroppstyp Primary
Värd Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunofluorescence Yes
Immunogen Human PSA
Rening purified by Protein A chromatography
Storage buffer PBS containing 0,09% sodium azide.
Förvaringstemperatur Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal