Anti-BAG6 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96553-50EA 0 SEK
BIRBORB96553-50
Anti-BAG6 Rabbit Polyclonal Antibody
Antikroppar
Immunochemistry: 5-10 ?g/ml; Western Blot: 1:5000-1:8000

Type: Primary
Antigen: BAG6 (BCL2-associated athanogene 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR