Anti-CDC42BPB Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96568-50EA 0 SEK
BIRBORB96568-50
Anti-CDC42BPB Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:40000; Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CDC42BPB (CDC42 binding protein kinase beta (DMPK-like))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR