Anti-ARRDC2 Rabbit Polyclonal Antibody

Leverantör: Biorbyt
BIRBORB96570-50EA 0 SEK
BIRBORB96570-50
Anti-ARRDC2 Rabbit Polyclonal Antibody
Antikroppar
ELISA: 1:40000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ARRDC2 (arrestin domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR