Anti-SLC22A1 Goat polyclonal antibody unconjugated
ABCAAB106947-100EA 7150 SEK
ABCAAB106947-100
Anti-SLC22A1 Goat polyclonal antibody unconjugated
Type: Primary
Antigen: SLC22A1 (solute carrier family 22 (organic cation transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human