Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: 25B881.1]

Leverantör: Novus Biologicals
NOVUNB100-56529EA 6825 SEK
NOVUNB100-56529
Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: 25B881.1]
Antikroppar
Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 25B881.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR