Anti-PCNA Mouse Monoclonal Antibody (DyLight® 405) [clone: PC10]

Leverantör: Novus Biologicals
NOVUNB500-106VEA 8400 SEK
NOVUNB500-106V
Anti-PCNA Mouse Monoclonal Antibody (DyLight® 405) [clone: PC10]
Antikroppar
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: PC10
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR