Anti-PKM Rabbit Polyclonal Antibody (Biotin)

Leverantör: Novus Biologicals
NOVUNBP1-48308BEA 8250 SEK
NOVUNBP1-48308B
Anti-PKM Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR