Anti-COPB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverantör: United States Biological
USBI034113-PEEA 9900 SEK
USBI034113-PE
Anti-COPB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: COPB1 (coatomer protein complex, subunit beta 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human