Vill du logga in igen?
Välkommen till en dag med Säkerhet i Fokus! – 20-23 maj – läs mer och anmäl dig nu!
Anti-COPB2 Rabbit Polyclonal Antibody
Leverantör: US Biological
USBI034114
Anti-COPB2 Rabbit Polyclonal Antibody
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: COPB2 (coatomer protein complex, subunit beta 2 (beta prime))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now
ORDER
SPECIFIKATIONER
RELATERAD INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal