Anti-COPS2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverantör: United States Biological
USBI034118-APEA 9900 SEK
USBI034118-AP
Anti-COPS2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: COPS2 (COP9 signalosome subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR