Anti-COPS2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverantör: United States Biological
USBI034118-APCEA 9900 SEK
USBI034118-APC
Anti-COPS2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: COPS2 (COP9 signalosome subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR