Anti-COQ7 Rabbit Polyclonal Antibody (Biotin)
Leverantör: United States Biological
USBI034126-BIOTINEA 9900 SEK
USBI034126-BIOTIN
Anti-COQ7 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Type: Primary
Antigen: COQ7 (coenzyme Q7 homolog, ubiquinone (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human