Anti-CORO6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: United States Biological
USBI034132-APCEA 9900 SEK
USBI034132-APC
Anti-CORO6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: CORO6 (coronin 6)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse