Anti-COX10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverantör: United States Biological
USBI034135-APCEA 9900 SEK
USBI034135-APC
Anti-COX10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikroppar
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: COX10 (cytochrome c oxidase assembly homolog 10 (yeast))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human