Anti-COX6B1 Rabbit Polyclonal Antibody

Leverantör: United States Biological
USBI034143EA 6825 SEK
USBI034143
Anti-COX6B1 Rabbit Polyclonal Antibody
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: COX6B1 (cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR