Anti-CPA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverantör: United States Biological
USBI034157-HRPEA 9900 SEK
USBI034157-HRP
Anti-CPA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: CPA2 (carboxypeptidase A2 (pancreatic))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human